Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-cyclopropyl-1H-pyrazole, 96%

$98.39 - $342.04

Chemical Identifiers

Chemical Name or Material 3-Amino-5-cyclopropyl-1H-pyrazole
CAS 175137-46-9
Molecular Formula C6H9N3
Formula Weight 123.16
MDL Number MFCD08461681
Synonym 5-Cyclopropyl-1H-pyrazole-3-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5089803

  SDS
alfa aesar™
H5089803
1g Each for $342.04

AAH50898MD

  SDS
alfa aesar™
H50898MD
250mg Each for $98.39
Description & Specifications

Specifications

Formula Weight 123.16
Chemical Name or Material 3-Amino-5-cyclopropyl-1H-pyrazole
Quantity 1g
Assay 96%
Synonym 5-Cyclopropyl-1H-pyrazole-3-amine
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C6H9N3
MDL Number MFCD08461681
CAS 175137-46-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.