Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-cyclobutyl-1H-pyrazole, 97%

$105.78 - $366.15

Chemical Identifiers

Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole
CAS 326827-21-8
Molecular Formula C7H11N3
Formula Weight 137.19
MDL Number MFCD08061045
Synonym 5-Cyclobutyl-1H-pyrazol-3-ylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5088003

  SDS
alfa aesar™
H5088003
1g Each for $366.15

AAH50880MD

  SDS
alfa aesar™
H50880MD
250mg Each for $105.78
Description & Specifications

Specifications

Formula Weight 137.19
Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole
Synonym 5-Cyclobutyl-1H-pyrazol-3-ylamine
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Quantity 1g
Assay 97%
Molecular Formula C7H11N3
MDL Number MFCD08061045
CAS 326827-21-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.