Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-chloropyridine, 95%

$272.73 - $1,011.04

Chemical Identifiers

Chemical Name or Material 3-Amino-5-chloropyridine
CAS 22353-34-0
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD03701386
Synonym 5-Chloro-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3183303

  SDS
alfa aesar™
H3183303
1g Each for $272.73

AAH3183306

  SDS
alfa aesar™
H3183306
5g Each for $1,011.04

AAH31833MD

  SDS
alfa aesar™
H31833MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-Amino-5-chloropyridine
CAS 22353-34-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 71° to 75°C
Quantity 1g
Odor Unpleasant
Assay 95%
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD03701386
Synonym 5-Chloro-3-pyridinamine
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.