missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-5-bromopyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441450010

 View more versions of this product

Catalog No. AC441450010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Amino-5-bromopyridine
Glass bottle
62.9°C to 65.2°C
13535-01-8
100.0
C5H5BrN2
MDQXGHBCDCOOSM-UHFFFAOYSA-N
5-bromopyridin-3-amine
817681
Powder
Authentic
Brown to White
1g
96.0
97%
3-amino-5-bromo pyridine, 3-amino-5-bromopyridine, 3-pyridinamine, 5-bromo, 5-bromo-3-aminopyridine, 5-bromo-3-pyridinamine, 5-bromo-3-pyridylamine, 5-bromo-pyridin-3-ylamine, 5-bromopyridin-3-ylamine, 5-brompyridin-3-amin, pubchem1213
C1=C(C=NC=C1Br)N
173.01
173.01
97%

Chemical Identifiers

13535-01-8
173.01
3-amino-5-bromo pyridine, 3-amino-5-bromopyridine, 3-pyridinamine, 5-bromo, 5-bromo-3-aminopyridine, 5-bromo-3-pyridinamine, 5-bromo-3-pyridylamine, 5-bromo-pyridin-3-ylamine, 5-bromopyridin-3-ylamine, 5-brompyridin-3-amin, pubchem1213
5-bromopyridin-3-amine
C5H5BrN2
MDQXGHBCDCOOSM-UHFFFAOYSA-N
817681
C1=C(C=NC=C1Br)N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents