missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-5-bromopyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 13535-01-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H5BrN2
Formula Weight 173.01
Synonym 5-Bromopyrid-3-ylamine
Chemical Name or Material 3-Amino-5-bromopyridine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441450010 SDS Thermo Fisher Scientific
AC441450010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441450100 Thermo Fisher Scientific
AC441450100
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13535-01-8
100.0
C5H5BrN2
5-Bromopyrid-3-ylamine
≥96.0%
96.0
97%
173.01
3-Amino-5-bromopyridine

Specifications

13535-01-8
100.0
Powder
White to yellow to brown
173.01
5-Bromopyrid-3-ylamine
≥96.0%
96.0
Authentic
97%
C5H5BrN2
1g
3-Amino-5-bromopyridine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.