Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-bromo-4-hydroxypyridine, 95%

Chemical Identifiers

Chemical Name or Material 3-Amino-5-bromo-4-hydroxypyridine
CAS 101084-20-2
Formula Weight 189.01
MDL Number MFCD08692340
Beilstein 121085
Synonym 3-Amino-5-bromo-4-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66810MD

 
alfa aesar™
H66810MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.01
CAS 101084-20-2
MDL Number MFCD08692340
Chemical Name or Material 3-Amino-5-bromo-4-hydroxypyridine
Physical Form Solid
Synonym 3-Amino-5-bromo-4-pyridinol
Beilstein 121085
Quantity 250mg
Storage Note 1 -30° to -10°C
Assay 95%
Safety and Handling

-30° to -10°C