missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-bromo-2-methylpyridine, 96%

Manufacturer:  Alfa Aesar™ H5466503

 View more versions of this product

Catalog No. AAH5466503



Specifications

Specifications

3-Amino-5-bromo-2-methylpyridine
1g
C6H7BrN2
3-amino-5-bromo-2-methylpyridine
CC1=NC=C(C=C1N)Br
187.04
187.04
108°C to 109°C
914358-73-9
MFCD09031418
CBAXYISWNGGXOZ-UHFFFAOYSA-N
5-bromo-2-methylpyridin-3-amine
21964096
96%

Chemical Identifiers

914358-73-9
187.04
CBAXYISWNGGXOZ-UHFFFAOYSA-N
21964096
CC1=NC=C(C=C1N)Br
C6H7BrN2
MFCD09031418
3-amino-5-bromo-2-methylpyridine
5-bromo-2-methylpyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents