missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-bromo-2-methoxypyridine, 96%

Manufacturer:  Alfa Aesar™ H5429906

 View more versions of this product

Catalog No. AAH5429906



Description

Description

Applications
3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Amino-5-bromo-2-methoxypyridine
5g
C6H7BrN2O
3-amino-5-bromo-2-methoxypyridine
COC1=NC=C(C=C1N)Br
203.039
203.04
53°C to 55°C
884495-39-0
MFCD07368887
HJOOFLFWIISCAI-UHFFFAOYSA-N
5-bromo-2-methoxypyridin-3-amine
44754869
96%

Chemical Identifiers

884495-39-0
203.039
HJOOFLFWIISCAI-UHFFFAOYSA-N
44754869
COC1=NC=C(C=C1N)Br
C6H7BrN2O
MFCD07368887
3-amino-5-bromo-2-methoxypyridine
5-bromo-2-methoxypyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents