missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-bromo-2-methoxypyridine, 96%

Manufacturer:  Alfa Aesar™ H5429903

 View more versions of this product

Catalog No. AAH5429903



Description

Description

Applications
3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Amino-5-bromo-2-methoxypyridine
1g
C6H7BrN2O
3-amino-5-bromo-2-methoxypyridine
HJOOFLFWIISCAI-UHFFFAOYSA-N
5-bromo-2-methoxypyridin-3-amine
44754869
96%
53°C to 55°C
884495-39-0
MFCD07368887
Slightly soluble in water.
COC1=NC=C(C=C1N)Br
203.039
203.04

Chemical Identifiers

884495-39-0
203.039
HJOOFLFWIISCAI-UHFFFAOYSA-N
44754869
COC1=NC=C(C=C1N)Br
C6H7BrN2O
MFCD07368887
3-amino-5-bromo-2-methoxypyridine
5-bromo-2-methoxypyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents