missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-bromo-2-hydroxypyridine, 95%

Manufacturer:  Alfa Aesar™ H61479MD

 View more versions of this product

Catalog No. AAH61479MD



Specifications

Specifications

3-Amino-5-bromo-2-hydroxypyridine
98786-86-8
MFCD03840438
ULOVLVWNSQNABA-UHFFFAOYSA-N
3-amino-5-bromo-1H-pyridin-2-one
13434515
Solid
250mg
C5H5BrN2O
2 1h-pyridinone, 3-amino-5-bromo
C1=C(C(=O)NC=C1Br)N
189.012
189.01
95%

Chemical Identifiers

98786-86-8
189.012
ULOVLVWNSQNABA-UHFFFAOYSA-N
13434515
C1=C(C(=O)NC=C1Br)N
C5H5BrN2O
MFCD03840438
2 1h-pyridinone, 3-amino-5-bromo
3-amino-5-bromo-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents