Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-bromo-2-chloropyridine, 97%

$61.88 - $224.73

Chemical Identifiers

Chemical Name or Material 3-Amino-5-bromo-2-chloropyridine
CAS 588729-99-1
Molecular Formula C5H4BrClN2
Formula Weight 207.46
MDL Number MFCD02682092
Synonym 5-Bromo-2-chloro-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3161403

  SDS
alfa aesar™
H3161403
1g Each for $61.88

AAH3161406

  SDS
alfa aesar™
H3161406
5g Each for $224.73
Description & Specifications

Specifications

Formula Weight 207.46
Chemical Name or Material 3-Amino-5-bromo-2-chloropyridine
Assay 97%
Melting Point 129° to 132°C
Molecular Formula C5H4BrClN2
Quantity 1g
MDL Number MFCD02682092
CAS 588729-99-1
Synonym 5-Bromo-2-chloro-3-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.