missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-5-bromo-1H-indazole, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435760010
Specifications
3-Amino-5-bromo-1H-indazole | |
Glass bottle | |
237°C to 238°C | |
61272-71-7 | |
100.0 | |
C7H6BrN3 | |
1h-indazol-3-amine, 5-bromo | |
C1=CC2=C(C=C1Br)C(=NN2)N | |
212.05 | |
212.05 | |
95% |
Authentic | |
Colorless | |
1g | |
94.0 | |
94% min. (HPLC) | |
MFCD03426696 | |
OMPYFDJVSAMSMA-UHFFFAOYSA-N | |
5-bromo-1H-indazol-3-amine | |
817910 | |
Liquid |
Chemical Identifiers
61272-71-7 | |
212.05 | |
OMPYFDJVSAMSMA-UHFFFAOYSA-N | |
817910 | |
C1=CC2=C(C=C1Br)C(=NN2)N |
C7H6BrN3 | |
MFCD03426696 | |
1h-indazol-3-amine, 5-bromo | |
5-bromo-1H-indazol-3-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning