Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5,6-dimethyl-1,2,4-triazine, 97%

$36.95 - $321.72

Chemical Identifiers

Chemical Name or Material 3-Amino-5,6-dimethyl-1,2,4-triazine
CAS 17584-12-2
Molecular Formula C5H8N4
Formula Weight 124.14
MDL Number MFCD00006460
Beilstein 118790
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1599306

 
alfa aesar™
A1599306
5g Each for $36.95

AAA1599314

 
alfa aesar™
A1599314
25g Each for $94.51

AAA1599322

 
alfa aesar™
A1599322
100g Each for $321.72
Description & Specifications

Specifications

Formula Weight 124.14
CAS 17584-12-2
Chemical Name or Material 3-Amino-5,6-dimethyl-1,2,4-triazine
Melting Point 210° to 212°C
Assay 97%
Quantity 5g
Molecular Formula C5H8N4
MDL Number MFCD00006460
Beilstein 118790
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XY3177100
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.