Learn More
3-Amino-5-(2-furyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals
$72.33 - $190.95
Chemical Identifiers
CAS | 96799-02-9 |
---|---|
Molecular Formula | C7H7N3O |
Molecular Weight (g/mol) | 149.153 |
MDL Number | MFCD00082696 |
InChI Key | XJNZHICOWTVWOX-UHFFFAOYSA-N |
Synonym | 3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine |
PubChem CID | 523183 |
IUPAC Name | 5-(furan-2-yl)-1H-pyrazol-3-amine |
SMILES | C1=COC(=C1)C2=CC(=NN2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL11721MD
|
Thermo Scientific Chemicals
L11721MD |
250 mg |
Each for $72.33
|
|
|||||
AAL1172103
|
Thermo Scientific Chemicals
L1172103 |
1 g |
Each for $190.95
|
|
|||||
Description
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
96799-02-9 | |
149.153 | |
XJNZHICOWTVWOX-UHFFFAOYSA-N | |
523183 | |
C1=COC(=C1)C2=CC(=NN2)N |
C7H7N3O | |
MFCD00082696 | |
3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine | |
5-(furan-2-yl)-1H-pyrazol-3-amine |
Specifications
96799-02-9 | |
C7H7N3O | |
250 mg | |
3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine | |
XJNZHICOWTVWOX-UHFFFAOYSA-N | |
5-(furan-2-yl)-1H-pyrazol-3-amine | |
523183 | |
97% |
124°C to 128°C | |
MFCD00082696 | |
4399373 | |
Insoluble in water. | |
C1=COC(=C1)C2=CC(=NN2)N | |
149.153 | |
149.15 | |
3-Amino-5-(2-furyl)-1H-pyrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only