missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-(2-furyl)-1H-pyrazole, 97%

Manufacturer:  Alfa Aesar™ L1172103

 View more versions of this product

Catalog No. AAL1172103



Description

Description

Applications
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

3-Amino-5-(2-furyl)-1H-pyrazole
1g
C7H7N3O
4399373
XJNZHICOWTVWOX-UHFFFAOYSA-N
5-(furan-2-yl)-1H-pyrazol-3-amine
523183
97%
124°C to 128°C
96799-02-9
MFCD00082696
3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine
C1=COC(=C1)C2=CC(=NN2)N
149.153
149.15

Chemical Identifiers

96799-02-9
149.153
XJNZHICOWTVWOX-UHFFFAOYSA-N
523183
C1=COC(=C1)C2=CC(=NN2)N
C7H7N3O
MFCD00082696
3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine
5-(furan-2-yl)-1H-pyrazol-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents