Learn More
3-Amino-4-methylpyridine, 98%, Thermo Scientific Chemicals
$130.14 - $361.38
Chemical Identifiers
CAS | 3430-27-1 |
---|---|
Molecular Formula | C6H8N2 |
Molecular Weight (g/mol) | 108.144 |
MDL Number | MFCD00128871 |
InChI Key | IBKMZYWDWWIWEL-UHFFFAOYSA-N |
Synonym | 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline |
PubChem CID | 137935 |
IUPAC Name | 4-methylpyridin-3-amine |
SMILES | CC1=C(C=NC=C1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1973006
|
Thermo Scientific Chemicals
L1973006 |
5 g |
Each for $130.14
|
|
|||||
AAL1973014
|
Thermo Scientific Chemicals
L1973014 |
25 g |
Each for $361.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3430-27-1 | |
108.144 | |
IBKMZYWDWWIWEL-UHFFFAOYSA-N | |
137935 | |
CC1=C(C=NC=C1)N |
C6H8N2 | |
MFCD00128871 | |
3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline | |
4-methylpyridin-3-amine |
Specifications
3430-27-1 | |
254°C | |
C6H8N2 | |
5 g | |
107792 | |
3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline | |
CC1=C(C=NC=C1)N | |
108.144 | |
108.14 | |
3-Amino-4-methylpyridine |
102°C to 106°C | |
Unpleasant | |
MFCD00128871 | |
UN2811 | |
Hygroscopic | |
IBKMZYWDWWIWEL-UHFFFAOYSA-N | |
4-methylpyridin-3-amine | |
137935 | |
98% |
Safety and Handling
GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only