Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-methylbiphenyl, 98%

$84.81 - $84.81

Chemical Identifiers

Chemical Name or Material 3-Amino-4-methylbiphenyl
CAS 80938-67-6
Molecular Formula C13H13N
Formula Weight 183.26
MDL Number MFCD00270126
Synonym 4-Methylbiphenyl-3-ylamine; 5-Phenyl-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2826503

  SDS
alfa aesar™
H2826503
1g Each for $84.81
Description & Specifications

Specifications

Formula Weight 183.26
Quantity 1g
Chemical Name or Material 3-Amino-4-methylbiphenyl
Assay 98%
Molecular Formula C13H13N
MDL Number MFCD00270126
CAS 80938-67-6
Synonym 4-Methylbiphenyl-3-ylamine; 5-Phenyl-o-toluidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.