missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-4-methylbenzoic Acid 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 103980500

 View more versions of this product

Catalog No. AC103980500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Amino-4-methylbenzoic acid
2458-12-0
98.5
Glass bottle
Pink to grayish
≥98.5%
99%
H2NC6H3(CH3)CO2H
MFCD00007742
99%
Authentic
100.0
164° to 168°C
Crystalline Powder
50g
C8H9NO2
151.16
14, 487

Chemical Identifiers

2458-12-0
100.0
C8H9NO2
151.16
3-Amino-4-methylbenzoic acid, 99%
98.5
99%
H2NC6H3(CH3)CO2H
MFCD00007742
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 219-543-1

TSCA : TSCA