Please login to your online account to display your discounted pricing

3-Amino-4-methylbenzoic Acid 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103982500

 View more versions of this product

Catalog No. AC103982500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 164° to 168°C
Color Pink to gray
Quantity 250g
Chemical Name or Material 3-Amino-4-methylbenzoic acid
Name Note 99%
CAS 2458-12-0
CAS Min % 98.5
CAS Max % 100.0
Physical Form Pink to grayish crystalline powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H9NO2
Linear Formula H2NC6H3(CH3)CO2H
Formula Weight 151.16g/mol
MDL Number MFCD00007742
Beilstein 14, 487
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 219-543-1
TSCA TSCA

Specifications

Chemical Name or Material 3-Amino-4-methylbenzoic acid
Name Note 99%
CAS 2458-12-0
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H9NO2
Linear Formula H2NC6H3(CH3)CO2H
Formula Weight 151.16g/mol
MDL Number MFCD00007742
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents