Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-methylbenzoic acid, 99%

$21.70 - $183.82

Chemical Identifiers

Chemical Name or Material 3-Amino-4-methylbenzoic acid
CAS 2458-12-0
Molecular Formula C8H9NO2
Formula Weight 151.16
MDL Number MFCD00007742
Beilstein 2082449
Synonym 3-Amino-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1267406

  SDS
alfa aesar™
A1267406
5g Each for $21.70

AAA1267422

  SDS
alfa aesar™
A1267422
100g Each for $183.82

AAA1267414

  SDS
alfa aesar™
A1267414
25g Each for $73.69
Description & Specifications

Specifications

Formula Weight 151.16
Molecular Formula C8H9NO2
Synonym 3-Amino-p-toluic acid
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 164° to 168°C
MDL Number MFCD00007742
CAS 2458-12-0
Health Hazard 1 Warning
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Amino-4-methylbenzoic acid
Beilstein 2082449

Applications
3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressures.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.