Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-methylbenzeneboronic acid, 98%

$108.34 - $372.18

Chemical Identifiers

Chemical Name or Material 3-Amino-4-methylbenzeneboronic acid
CAS 22237-12-3
Molecular Formula C7H10BNO2
Formula Weight 150.98
MDL Number MFCD01074640
Synonym 3-Amino-4-methylphenylboronic acid; 3-Amino-p-tolylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1769503

  SDS
alfa aesar™
L1769503
1g Each for $108.34

AAL1769506

  SDS
alfa aesar™
L1769506
5g Each for $372.18
Description & Specifications

Specifications

Formula Weight 150.98
MDL Number MFCD01074640
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Amino-4-methylbenzeneboronic acid
CAS 22237-12-3
Quantity 1g
Synonym 3-Amino-4-methylphenylboronic acid; 3-Amino-p-tolylboronic acid
Melting Point ≈250°C
Assay 98%
Health Hazard 1 Warning
Molecular Formula C7H10BNO2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.