missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-4-methylbenzamide, 98%, ACROS Organics™

Chemical Identifiers

CAS 19406-86-1
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00035936
InChI Key VYBKAZXQKUFAHG-UHFFFAOYSA-N
Synonym 3-amino-4-methyl benzamide, 3-amino-4-methyl-benzamide, 3-amino-p-toluamide, 3-azanyl-4-methyl-benzamide, acmc-1c8qe, atad, benzamide, 3-amino-4-methyl, dsstox_cid_24407, dsstox_rid_80206, unii-51vb4677ff
PubChem CID 88043
IUPAC Name 3-amino-4-methylbenzamide
SMILES CC1=C(C=C(C=C1)C(=O)N)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC357050050
SDS View Documents
Thermo Fisher Scientific
357050050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

19406-86-1
150.18
VYBKAZXQKUFAHG-UHFFFAOYSA-N
88043
CC1=C(C=C(C=C1)C(=O)N)N
C8H10N2O
MFCD00035936
3-amino-4-methyl benzamide, 3-amino-4-methyl-benzamide, 3-amino-p-toluamide, 3-azanyl-4-methyl-benzamide, acmc-1c8qe, atad, benzamide, 3-amino-4-methyl, dsstox_cid_24407, dsstox_rid_80206, unii-51vb4677ff
3-amino-4-methylbenzamide

Specifications

Authentic
Brown
5g
97.5
97.5% min. (GC)
NH2C6H3(CH3)CONH2
3-amino-4-methyl benzamide, 3-amino-4-methyl-benzamide, 3-amino-p-toluamide, 3-azanyl-4-methyl-benzamide, acmc-1c8qe, atad, benzamide, 3-amino-4-methyl, dsstox_cid_24407, dsstox_rid_80206, unii-51vb4677ff
CC1=C(C=C(C=C1)C(=O)N)N
150.18
150.18
98%
Glass Bottle
128.0°C to 132.0°C
19406-86-1
100.0
C8H10N2O
MFCD00035936
VYBKAZXQKUFAHG-UHFFFAOYSA-N
3-amino-4-methylbenzamide
88043
Crystalline Chunks
3-Amino-4-methylbenzamide, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-039-0

TSCA : TSCA

SDS
Product Certifications