Please login to your online account to display your discounted pricing

3-Amino-4-methylbenzamide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  357050050

 View more versions of this product

Catalog No. AC357050050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Chunks
Assay ≥97.5%
Formula Weight 150.18
Quantity 5g
Color Light brown
TSCA TSCA
EINECS Number 243-039-0
Melting Point 128° to 132°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H10N2O
MDL Number MFCD00035936
Chemical Name or Material 3-Amino-4-methylbenzamide
Name Note 98%
Assay Percent Range 98%
CAS Min % 97.5
Linear Formula NH2C6H3(CH3)CONH2
CAS 19406-86-1

Specifications

Chemical Name or Material 3-Amino-4-methylbenzamide
Name Note 98%
CAS 19406-86-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10N2O
Linear Formula NH2C6H3(CH3)CONH2
Formula Weight 150.18
MDL Number MFCD00035936
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents