Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-methylbenzamide, 97%

$266.70 - $266.70

Chemical Identifiers

Chemical Name or Material 3-Amino-4-methylbenzamide
CAS 19406-86-1
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00035936
Synonym 3-Amino-p-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2536714

  SDS
alfa aesar™
B2536714
25g Each for $266.70
Description & Specifications

Specifications

Formula Weight 150.18
MDL Number MFCD00035936
Chemical Name or Material 3-Amino-4-methylbenzamide
Assay 97%
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Molecular Formula C8H10N2O
Synonym 3-Amino-p-toluamide
Melting Point 127° to 131°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 19406-86-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.