Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-methoxybenzanilide, 98%

$74.45 - $74.45

Chemical Identifiers

Chemical Name or Material 3-Amino-4-methoxybenzanilide
CAS 120-35-4
Molecular Formula C14H14N2O2
Formula Weight 242.28
MDL Number MFCD00017166
Synonym 3-Amino-p-anisanilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1916809

  SDS
alfa aesar™
A1916809
10g Each for $74.45
Description & Specifications

Specifications

Formula Weight 242.28
Molecular Formula C14H14N2O2
CAS 120-35-4
RTECS Number CV7400000
MDL Number MFCD00017166
Quantity 10g
TSCA TSCA
Synonym 3-Amino-p-anisanilide
Assay 98%
Chemical Name or Material 3-Amino-4-methoxybenzanilide
Melting Point 154° to 156°C