Please login to your online account to display your discounted pricing

3-Amino-4-iodo-1H-indazole, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Amino-4-iodo-1H-indazole
CAS 599191-73-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C7H6IN3
Formula Weight 259.05
MDL Number MFCD03426697
Synonym 4-Iodo-1H-indazol-3-ylamine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC435750050

  SDS
thermo fisher scientific
AC435750050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94.0%
Physical Form Crystalline Solid
Formula Weight 259.05
Quantity 5g
Color Off-white to brown
Melting Point 150° to 155°C
CAS Min % 94.0
CAS Max % 100.0
Synonym 4-Iodo-1H-indazol-3-ylamine
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD03426697
CAS 599191-73-8
Molecular Formula C7H6IN3
Assay Percent Range 95%
Chemical Name or Material 3-Amino-4-iodo-1H-indazole
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.