Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-hydroxypyridine hydrate, 95%

$105.03 - $105.03

Chemical Identifiers

Chemical Name or Material 3-Amino-4-hydroxypyridine hydrate
CAS 6320-39-4
Molecular Formula C5H6N2O·xH2O
Formula Weight 110.12 (Anhydrous)
MDL Number MFCD04114230
Synonym 3-Amino-4-pyridinol; 3-Amino-4(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2720503

  SDS
alfa aesar™
H2720503
1g Each for $105.03
Description & Specifications

Specifications

Formula Weight 110.12 (Anhydrous)
MDL Number MFCD04114230
Chemical Name or Material 3-Amino-4-hydroxypyridine hydrate
Molecular Formula C5H6N2O·xH2O
Melting Point 145° to 149°C
CAS 6320-39-4
Health Hazard 1 Warning
Synonym 3-Amino-4-pyridinol; 3-Amino-4(1H)-pyridone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.