missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-4-hydroxyphenylboronic acid pinacol ester, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444510050
Specifications
3-Amino-4-hydroxyphenylboronic acid pinacol ester | |
5g | |
95% | |
2-amino-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-amino-4-hydroxyphenylboronic acid pinacol ester, 3-amino-4-hydroxyphenylboronic acid, pinacol ester, phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)N | |
235.09 | |
235.09 |
151°C to 152°C | |
760990-10-1 | |
C12H18BNO3 | |
RXHWPAXQYXTPJB-UHFFFAOYSA-N | |
2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
46738034 | |
95% |
Chemical Identifiers
760990-10-1 | |
235.09 | |
2-amino-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-amino-4-hydroxyphenylboronic acid pinacol ester, 3-amino-4-hydroxyphenylboronic acid, pinacol ester, phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
C12H18BNO3 | |
RXHWPAXQYXTPJB-UHFFFAOYSA-N | |
46738034 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning