Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-hydroxybenzoic acid, 97%

$44.89 - $623.80

Chemical Identifiers

Chemical Name or Material 3-Amino-4-hydroxybenzoic acid
CAS 1571-72-8
Molecular Formula C7H7NO3
Formula Weight 153.14
MDL Number MFCD00007697
Beilstein 972238
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1348706

  SDS
alfa aesar™
A1348706
5g Each for $44.89

AAA1348714

  SDS
alfa aesar™
A1348714
25g Each for $180.81

AAA1348722

  SDS
alfa aesar™
A1348722
100g Each for $623.80
Description & Specifications

Specifications

Formula Weight 153.14
Chemical Name or Material 3-Amino-4-hydroxybenzoic acid
Assay 97%
Molecular Formula C7H7NO3
Melting Point ≈200°C (dec.)
MDL Number MFCD00007697
CAS 1571-72-8
Quantity 5g
Beilstein 972238
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Amino-4-hydroxybenzoic acid is used to produce 4-acetoxy-3-acetylamino-benzoic acid with acetic acid anhydride at temperature of 0 - 20°C. This reaction will need reagents aq. HCl and NaOAc.

Solubility
Soluble in solvents.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.