Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-hydroxybenzhydrazide, 98%

$54.85 - $926.66

Chemical Identifiers

Chemical Name or Material 3-Amino-4-hydroxybenzhydrazide
CAS 7450-57-9
Molecular Formula C7H9N3O2
Formula Weight 167.17
MDL Number MFCD00017068
Beilstein 6503217
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1153803

  SDS
alfa aesar™
A1153803
1g Each for $54.85

AAA1153806

  SDS
alfa aesar™
A1153806
5g Each for $209.45

AAA1153814

  SDS
alfa aesar™
A1153814
25g Each for $926.66
Description & Specifications

Specifications

Formula Weight 167.17
Molecular Formula C7H9N3O2
MDL Number MFCD00017068
CAS 7450-57-9
Beilstein 6503217
Health Hazard 1 Warning
Quantity 1g
Assay 98%
Melting Point 250° to 252°C
Chemical Name or Material 3-Amino-4-hydroxybenzhydrazide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.