missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-fluorobenzeneboronic acid pinacol ester, 96%

$69.11 - $221.80

Chemical Identifiers

CAS 1003575-43-6
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.081
MDL Number MFCD11044430
InChI Key VXNIVUKZNDRJBW-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine
PubChem CID 46737990
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62228MD View Documents Alfa Aesar™
H62228MD
250mg Each for $69.11
 
AAH6222803 View Documents Alfa Aesar™
H6222803
1g Each for $221.80
 
Specifications

Chemical Identifiers

1003575-43-6
237.081
VXNIVUKZNDRJBW-UHFFFAOYSA-N
46737990
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)N
C12H17BFNO2
MFCD11044430
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

Specifications

250mg
C12H17BFNO2
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)N
237.081
237.08
96%
1003575-43-6
MFCD11044430
VXNIVUKZNDRJBW-UHFFFAOYSA-N
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
46737990
Solid
3-Amino-4-fluorobenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures