missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6222803

 View more versions of this product

Catalog No. AAH6222803



Specifications

Specifications

3-Amino-4-fluorobenzeneboronic acid pinacol ester
1003575-43-6
MFCD11044430
VXNIVUKZNDRJBW-UHFFFAOYSA-N
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
46737990
Solid
1g
C12H17BFNO2
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)N
237.081
237.08
96%

Chemical Identifiers

1003575-43-6
237.081
VXNIVUKZNDRJBW-UHFFFAOYSA-N
46737990
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)N
C12H17BFNO2
MFCD11044430
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents