Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-chloropyridine, 98+%

$116.92 - $375.19

Chemical Identifiers

Chemical Name or Material 3-Amino-4-chloropyridine
CAS 20511-15-3
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD00235124
Beilstein 110187
Synonym 4-Chloro-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1972603

  SDS
alfa aesar™
L1972603
1g Each for $116.92

AAL1972606

  SDS
alfa aesar™
L1972606
5g Each for $375.19
Description & Specifications

Specifications

Formula Weight 128.56
MDL Number MFCD00235124
CAS 20511-15-3
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay ≥98%
Synonym 4-Chloro-3-pyridinamine
Beilstein 110187
Health Hazard 1 Warning
Chemical Name or Material 3-Amino-4-chloropyridine
Odor Unpleasant
Quantity 1g
Molecular Formula C5H5ClN2
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 59° to 62°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.