missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-chlorophenol, 97%

$70.24 - $901.64

Chemical Identifiers

CAS 16026-77-0
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD03427127
InChI Key FWUALUHYKLDYAN-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxyaniline
PubChem CID 5070078
IUPAC Name 3-amino-4-chlorophenol
SMILES C1=CC(=C(C=C1O)N)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5582603 View Documents Alfa Aesar™
H5582603
1g Each for $70.24
 
AAH5582606 View Documents Alfa Aesar™
H5582606
5g Each for $253.93
 
AAH5582614 View Documents Alfa Aesar™
H5582614
25g Each for $901.64
 
Description

Description

Applications
3-Amino-4-chlorophenol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic chemical intermediate for synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

16026-77-0
143.57
FWUALUHYKLDYAN-UHFFFAOYSA-N
5070078
C1=CC(=C(C=C1O)N)Cl
C6H6ClNO
MFCD03427127
2-chloro-5-hydroxyaniline
3-amino-4-chlorophenol

Specifications

Brown to Orange
1g
C6H6ClNO
2-chloro-5-hydroxyaniline
FWUALUHYKLDYAN-UHFFFAOYSA-N
3-amino-4-chlorophenol
5070078
Powder
3-Amino-4-chlorophenol
151°C to 154°C
16026-77-0
MFCD03427127
Slightly soluble in water.
C1=CC(=C(C=C1O)N)Cl
143.57
143.57
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures