missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-chlorophenol, 97%

Manufacturer:  Alfa Aesar™ H5582606

 View more versions of this product

Catalog No. AAH5582606



Description

Description

Applications
3-Amino-4-chlorophenol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic chemical intermediate for synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

3-Amino-4-chlorophenol
151°C to 154°C
16026-77-0
MFCD03427127
Slightly soluble in water.
C1=CC(=C(C=C1O)N)Cl
143.57
143.57
97%
Brown to Orange
5g
C6H6ClNO
2-chloro-5-hydroxyaniline, 3-amino-4-chlorophenol, 3-azanyl-4-chloranyl-phenol, 4-chloro-3-aminophenol, acmc-1bo5g, phenol, 3-amino-4-chloro, phenol,3-amino-4-chloro
FWUALUHYKLDYAN-UHFFFAOYSA-N
3-amino-4-chlorophenol
5070078
Powder

Chemical Identifiers

16026-77-0
143.57
FWUALUHYKLDYAN-UHFFFAOYSA-N
5070078
C1=CC(=C(C=C1O)N)Cl
C6H6ClNO
MFCD03427127
2-chloro-5-hydroxyaniline, 3-amino-4-chlorophenol, 3-azanyl-4-chloranyl-phenol, 4-chloro-3-aminophenol, acmc-1bo5g, phenol, 3-amino-4-chloro, phenol,3-amino-4-chloro
3-amino-4-chlorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents