Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4-chlorobenzoic acid, 98%

$895.86 - $895.86

Chemical Identifiers

Chemical Name or Material 3-Amino-4-chlorobenzoic acid
CAS 2840-28-0
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00007671
Beilstein 2085846
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1267136

  SDS
alfa aesar™
A1267136
500g Each for $895.86
Description & Specifications

Specifications

Formula Weight 171.58
Chemical Name or Material 3-Amino-4-chlorobenzoic acid
Quantity 500g
Molecular Formula C7H6ClNO2
TSCA TSCA
Melting Point 213° to 218°C
Assay 98%
MDL Number MFCD00007671
CAS 2840-28-0
Beilstein 2085846
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.