missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-4-carbethoxypyrazole, 99%, ACROS Organics™
$113.05 - $113.05
Chemical Identifiers
CAS | 6994-25-8 |
---|---|
Molecular Formula | C6H9N3O2 |
Molecular Weight (g/mol) | 155.16 |
MDL Number | MFCD00005238 |
InChI Key | YPXGHKWOJXQLQU-UHFFFAOYSA-N |
Synonym | 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-4-carbethoxypyrazol, 3-amino-4-carbethoxypyrazole, 3-amino-4-ethoxycarbonylpyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, ethyl 3-aminopyrazole-4-carboxylate, unii-9gy5ha1gaa |
PubChem CID | 81472 |
IUPAC Name | ethyl 5-amino-1H-pyrazole-4-carboxylate |
SMILES | CCOC(=O)C1=C(NN=C1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC103310050
|
Thermo Fisher Scientific
103310050 |
5g |
Each for $113.05
|
|
Chemical Identifiers
6994-25-8 | |
155.16 | |
YPXGHKWOJXQLQU-UHFFFAOYSA-N | |
81472 | |
CCOC(=O)C1=C(NN=C1)N |
C6H9N3O2 | |
MFCD00005238 | |
1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-4-carbethoxypyrazol, 3-amino-4-carbethoxypyrazole, 3-amino-4-ethoxycarbonylpyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, ethyl 3-aminopyrazole-4-carboxylate, unii-9gy5ha1gaa | |
ethyl 5-amino-1H-pyrazole-4-carboxylate |
Specifications
Authentic | |
White to Yellow | |
5g | |
98.5 | |
98.5% min. (HPLC) | |
MFCD00005238 | |
YPXGHKWOJXQLQU-UHFFFAOYSA-N | |
ethyl 5-amino-1H-pyrazole-4-carboxylate | |
81472 | |
Crystalline Powder | |
3-Amino-4-carbethoxypyrazole, 99% |
Glass bottle | |
103.0°C to 107.0°C | |
6994-25-8 | |
100.0 | |
C6H9N3O2 | |
1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-4-carbethoxypyrazol, 3-amino-4-carbethoxypyrazole, 3-amino-4-ethoxycarbonylpyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, ethyl 3-aminopyrazole-4-carboxylate, unii-9gy5ha1gaa | |
CCOC(=O)C1=C(NN=C1)N | |
155.16 | |
155.16 | |
99% |
Safety and Handling
EINECSNumber : 230-262-3