missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-4,4,4-trifluorobutyric acid hydrochloride, 95%

$99.01 - $337.73

Chemical Identifiers

CAS 91291-66-6
Molecular Formula C4H7ClF3NO2
Molecular Weight (g/mol) 193.55
MDL Number MFCD00181951
InChI Key MRMARVCQZOYQPI-UHFFFAOYSA-N
Synonym 3-amino-4,4,4-trifluorobutanoic acid hydrochloride, 3-amino-4,4,4-trifluorobutyric acid hydrochloride
PubChem CID 14442472
IUPAC Name 3-amino-4,4,4-trifluorobutanoic acid;hydrochloride
SMILES C(C(C(F)(F)F)N)C(=O)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL18336MD View Documents Alfa Aesar™
L18336MD
250mg Each for $99.01
 
AAL1833603 View Documents Alfa Aesar™
L1833603
1g Each for $337.73
 
Specifications

Chemical Identifiers

91291-66-6
193.55
MRMARVCQZOYQPI-UHFFFAOYSA-N
14442472
C(C(C(F)(F)F)N)C(=O)O.Cl
C4H7ClF3NO2
MFCD00181951
3-amino-4,4,4-trifluorobutanoic acid hydrochloride, 3-amino-4,4,4-trifluorobutyric acid hydrochloride
3-amino-4,4,4-trifluorobutanoic acid;hydrochloride

Specifications

197°C to 199°C
91291-66-6
MFCD00181951
MRMARVCQZOYQPI-UHFFFAOYSA-N
3-amino-4,4,4-trifluorobutanoic acid;hydrochloride
14442472
95%
250mg
C4H7ClF3NO2
3-amino-4,4,4-trifluorobutanoic acid hydrochloride, 3-amino-4,4,4-trifluorobutyric acid hydrochloride
C(C(C(F)(F)F)N)C(=O)O.Cl
193.55
193.55
3-Amino-4,4,4-trifluorobutyric acid hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning