Please login to your online account to display your discounted pricing

3-Amino-3-(p-chlorophenyl)propionic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334330050

 View more versions of this product

Catalog No. AC334330050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 199.64
Quantity 5g
Color Off-white
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C9H10ClNO2
MDL Number MFCD00126370
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material 3-Amino-3-(p-chlorophenyl)propionic acid
Linear Formula ClC6H4CH(NH2)CH2COOH
CAS 19947-39-8
CAS Min % 97.5
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Amino-3-(p-chlorophenyl)propionic acid
Name Note 98%
CAS 19947-39-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10ClNO2
Linear Formula ClC6H4CH(NH2)CH2COOH
Formula Weight 199.64
MDL Number MFCD00126370