missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-3-(p-chlorophenyl)propionic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 19947-39-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.64
MDL Number MFCD00126370
InChI Key BXGDBHAMTMMNTO-UHFFFAOYSA-N
Synonym 3-amino-3-4-chloro-phenyl-propionic acid, 3-amino-3-4-chlorophenyl propanoic acid, 3-amino-3-4-chlorophenyl propionic acid, 3-amino-3-p-chlorophenyl propionic acid, 3-p-chlorophenyl-dl-beta-alanine, dl-3-amino-3-4'-chlorophenyl propionic acid, dl-3-amino-3-4-chlorophenyl-propionic acid, dl-beta-4-chlorophenyl alanine, dl-beta-p-chlorophenyl alanine, rac-3-amino-3-4-chlorophenyl propionic acid
PubChem CID 344176
IUPAC Name 3-amino-3-(4-chlorophenyl)propanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)N)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334330010
SDS View Documents
Thermo Fisher Scientific
334330010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC334330050
SDS View Documents
Thermo Fisher Scientific
334330050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

19947-39-8
199.64
BXGDBHAMTMMNTO-UHFFFAOYSA-N
344176
C1=CC(=CC=C1C(CC(=O)O)N)Cl
C9H10ClNO2
MFCD00126370
3-amino-3-4-chloro-phenyl-propionic acid, 3-amino-3-4-chlorophenyl propanoic acid, 3-amino-3-4-chlorophenyl propionic acid, 3-amino-3-p-chlorophenyl propionic acid, 3-p-chlorophenyl-dl-beta-alanine, dl-3-amino-3-4'-chlorophenyl propionic acid, dl-3-amino-3-4-chlorophenyl-propionic acid, dl-beta-4-chlorophenyl alanine, dl-beta-p-chlorophenyl alanine, rac-3-amino-3-4-chlorophenyl propionic acid
3-amino-3-(4-chlorophenyl)propanoic acid

Specifications

Authentic
White
1g
97.5
98%
ClC6H4CH(NH2)CH2CO2H
3-amino-3-4-chloro-phenyl-propionic acid, 3-amino-3-4-chlorophenyl propanoic acid, 3-amino-3-4-chlorophenyl propionic acid, 3-amino-3-p-chlorophenyl propionic acid, 3-p-chlorophenyl-dl-beta-alanine, dl-3-amino-3-4'-chlorophenyl propionic acid, dl-3-amino-3-4-chlorophenyl-propionic acid, dl-beta-4-chlorophenyl alanine, dl-beta-p-chlorophenyl alanine, rac-3-amino-3-4-chlorophenyl propionic acid
C1=CC(=CC=C1C(CC(=O)O)N)Cl
199.64
199.64
98%
Glass bottle
243°C to 245°C
19947-39-8
100.0
C9H10ClNO2
MFCD00126370
BXGDBHAMTMMNTO-UHFFFAOYSA-N
3-amino-3-(4-chlorophenyl)propanoic acid
344176
Powder
3-Amino-3-(p-chlorophenyl)propionic acid, 98%
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
Product Certifications