Learn More
3-Amino-3-methyloxetane, 95%, Thermo Scientific Chemicals
$504.42 - $836.78
Chemical Identifiers
CAS | 874473-14-0 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.122 |
MDL Number | MFCD11111713 |
InChI Key | NQVWMPOQWBDSAI-UHFFFAOYSA-N |
Synonym | 3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane |
PubChem CID | 46835725 |
IUPAC Name | 3-methyloxetan-3-amine |
SMILES | CC1(COC1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57588MD
|
Thermo Scientific Chemicals
H57588MD |
250 mg |
Each for $504.42
|
|
|||||
AAH57588ME
|
Thermo Scientific Chemicals
H57588ME |
500 mg |
Each for $836.78
|
|
|||||
Description
It is used as an OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an OLED intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
874473-14-0 | |
87.122 | |
NQVWMPOQWBDSAI-UHFFFAOYSA-N | |
46835725 | |
CC1(COC1)N |
C4H9NO | |
MFCD11111713 | |
3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane | |
3-methyloxetan-3-amine |
Specifications
874473-14-0 | |
102°C | |
MFCD11111713 | |
3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane | |
NQVWMPOQWBDSAI-UHFFFAOYSA-N | |
3-methyloxetan-3-amine | |
46835725 | |
95% | |
3-Amino-3-methyloxetane |
0.99 g/mL | |
C4H9NO | |
250 mg | |
Slightly soluble in water. | |
CC1(COC1)N | |
87.122 | |
87.12 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only