Please login to your online account to display your discounted pricing

3-Amino-3-cyclohexylpropionic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334350050

 View more versions of this product

Catalog No. AC334350050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Flakes
Formula Weight 171.24
Quantity 5g
Color White
TSCA TSCA
CAS Max % 100.0
MDL Number MFCD01863250
Assay Percent Range 98%
Linear Formula C6H11CH(NH2)CH2COOH
CAS Min % 97.5
Chemical Name or Material 3-Amino-3-cyclohexylpropionic acid
Name Note 98%
Packaging Glass bottle
Molecular Formula C9 H17 N O2
CAS 129042-71-3
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Amino-3-cyclohexylpropionic acid
Name Note 98%
CAS 129042-71-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H17 N O2
Linear Formula C6H11CH(NH2)CH2COOH
Formula Weight 171.24
MDL Number MFCD01863250