Please login to your online account to display your discounted pricing

3-Amino-3-cyclohexylpropionic acid, 98%, ACROS Organics™

$1,039.18 - $1,039.18

Chemical Identifiers

Chemical Name or Material 3-Amino-3-cyclohexylpropionic acid
Name Note 98%
CAS 129042-71-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H17 N O2
Linear Formula C6H11CH(NH2)CH2COOH
Formula Weight 171.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334350050

  SDS
thermo fisher scientific
334350050
Glass bottle 5g Each for $1,039.18
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals or Flakes
Formula Weight 171.24
Quantity 5g
Color White
TSCA TSCA
CAS Max % 100.0
MDL Number MFCD01863250
Assay Percent Range 98%
Linear Formula C6H11CH(NH2)CH2COOH
CAS Min % 97.5
Chemical Name or Material 3-Amino-3-cyclohexylpropionic acid
Name Note 98%
Packaging Glass bottle
Molecular Formula C9 H17 N O2
CAS 129042-71-3
Infrared Spectrum Authentic