Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-3-(4-fluorophenyl)propionic acid, 96%

$64.34 - $64.34

Chemical Identifiers

Chemical Name or Material 3-Amino-3-(4-fluorophenyl)propionic acid
CAS 325-89-3
Molecular Formula C9H10FNO2
Formula Weight 183.18
MDL Number MFCD00181811
Beilstein 3258498
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19799MD

  SDS
alfa aesar™
L19799MD
250mg Each for $64.34
Description & Specifications

Specifications

Formula Weight 183.18
MDL Number MFCD00181811
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
CAS 325-89-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Molecular Formula C9H10FNO2
Health Hazard 1 Warning
Beilstein 3258498
Melting Point 214°C
Quantity 250mg
Chemical Name or Material 3-Amino-3-(4-fluorophenyl)propionic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C