Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-3-(4-chlorophenyl)propionic acid, 97%

$117.38 - $117.38

Chemical Identifiers

Chemical Name or Material 3-Amino-3-(4-chlorophenyl)propionic acid
CAS 19947-39-8
Molecular Formula C9H10ClNO2
Formula Weight 199.64
MDL Number MFCD00126370
Beilstein 2365902
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1979803

  SDS
alfa aesar™
L1979803
1g Each for $117.38
Description & Specifications

Specifications

Formula Weight 199.64
MDL Number MFCD00126370
Recommended Storage 2° to 8°C
CAS 19947-39-8
Assay 97%
Beilstein 2365902
Melting Point 214° to 216°C
Quantity 1g
Molecular Formula C9H10ClNO2
Chemical Name or Material 3-Amino-3-(4-chlorophenyl)propionic acid
Safety and Handling

2° to 8°C