Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-3-(3-methoxyphenyl)propionic acid, 97%

$44.45 - $119.78

Chemical Identifiers

Chemical Name or Material 3-Amino-3-(3-methoxyphenyl)propionic acid
CAS 68208-19-5
Molecular Formula C10H13NO3
Formula Weight 195.22
MDL Number MFCD00462569
Beilstein 2838792
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1980003

  SDS
alfa aesar™
L1980003
1g Each for $119.78

AAL19800MD

  SDS
alfa aesar™
L19800MD
250mg Each for $44.45
Description & Specifications

Specifications

Formula Weight 195.22
Chemical Name or Material 3-Amino-3-(3-methoxyphenyl)propionic acid
Molecular Formula C10H13NO3
Assay 97%
CAS 68208-19-5
MDL Number MFCD00462569
Recommended Storage 2° to 8°C
Beilstein 2838792
Quantity 1g
Melting Point 208°C
Safety and Handling

2° to 8°C