Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-3-(3-chlorophenyl)propionic acid, 98%

$76.05 - $76.05

Chemical Identifiers

Chemical Name or Material 3-Amino-3-(3-chlorophenyl)propionic acid
CAS 68208-21-9
Molecular Formula C9H10ClNO2
Formula Weight 199.64
MDL Number MFCD00463160
Beilstein 2938807
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19797MD

 
alfa aesar™
L19797MD
250mg Each for $76.05
Description & Specifications

Specifications

Formula Weight 199.64
Molecular Formula C9H10ClNO2
MDL Number MFCD00463160
CAS 68208-21-9
Chemical Name or Material 3-Amino-3-(3-chlorophenyl)propionic acid
Melting Point ≈217°C
Quantity 250mg
Assay 98%
Beilstein 2938807
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C