Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-3-(2-chlorophenyl)propionic acid, 97%

$53.42 - $118.07

Chemical Identifiers

Chemical Name or Material 3-Amino-3-(2-chlorophenyl)propionic acid
CAS 68208-20-8
Molecular Formula C9H10ClNO2
Formula Weight 199.64
MDL Number MFCD01871299
Beilstein 2938797
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19796MD

 
alfa aesar™
L19796MD
250mg Each for $53.42

AAL1979603

 
alfa aesar™
L1979603
1g Each for $118.07
Description & Specifications

Specifications

Formula Weight 199.64
Chemical Name or Material 3-Amino-3-(2-chlorophenyl)propionic acid
Assay 97%
MDL Number MFCD01871299
CAS 68208-20-8
Melting Point ≈223°C
Beilstein 2938797
Recommended Storage 2° to 8°C
Quantity 250mg
Molecular Formula C9H10ClNO2
Safety and Handling

2° to 8°C