Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-pyrazoleethanol, 96%

Chemical Identifiers

Chemical Name or Material 3-Amino-2-pyrazoleethanol
CAS 73616-27-0
Formula Weight 127.15
MDL Number MFCD08063809
Beilstein 607610
Synonym 5-Amino-1-(2-hydroxyethyl)pyrazole; 3-Aminopyrazole-2-ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6606606

 
alfa aesar™
H6606606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6606614

 
alfa aesar™
H6606614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6606622

 
alfa aesar™
H6606622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.15
Chemical Name or Material 3-Amino-2-pyrazoleethanol
CAS 73616-27-0
Assay 96%
MDL Number MFCD08063809
Synonym 5-Amino-1-(2-hydroxyethyl)pyrazole; 3-Aminopyrazole-2-ethanol
Physical Form Solid
Quantity 5g
Beilstein 607610
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C