Learn More
3-Amino-2-nitropyridine, 97%, Thermo Scientific Chemicals
$359.55 - $359.55
Chemical Identifiers
CAS | 13269-19-7 |
---|---|
Molecular Formula | C5H5N3O2 |
Molecular Weight (g/mol) | 139.114 |
MDL Number | MFCD00044102 |
InChI Key | GZBKVUGZEAJYHH-UHFFFAOYSA-N |
Synonym | 3-amino-2-nitropyridine, 2-nitro-3-pyridinamine, 2-nitro-3-aminopyridine, 3-pyridinamine, 2-nitro, 2-nitropyridin-3-amin, aminonitropyridine, amino nitro pyridine, pubchem2087, acmc-209bps, 2-nitro-3-pyridylamine |
PubChem CID | 83281 |
IUPAC Name | 2-nitropyridin-3-amine |
SMILES | C1=CC(=C(N=C1)[N+](=O)[O-])N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1961906
|
Thermo Scientific Chemicals
L1961906 |
5 g |
Each for $359.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13269-19-7 | |
139.114 | |
GZBKVUGZEAJYHH-UHFFFAOYSA-N | |
83281 | |
C1=CC(=C(N=C1)[N+](=O)[O-])N |
C5H5N3O2 | |
MFCD00044102 | |
3-amino-2-nitropyridine, 2-nitro-3-pyridinamine, 2-nitro-3-aminopyridine, 3-pyridinamine, 2-nitro, 2-nitropyridin-3-amin, aminonitropyridine, amino nitro pyridine, pubchem2087, acmc-209bps, 2-nitro-3-pyridylamine | |
2-nitropyridin-3-amine |
Specifications
13269-19-7 | |
C5H5N3O2 | |
5 g | |
3-amino-2-nitropyridine, 2-nitro-3-pyridinamine, 2-nitro-3-aminopyridine, 3-pyridinamine, 2-nitro, 2-nitropyridin-3-amin, aminonitropyridine, amino nitro pyridine, pubchem2087, acmc-209bps, 2-nitro-3-pyridylamine | |
C1=CC(=C(N=C1)[N+](=O)[O-])N | |
139.114 | |
139.11 | |
3-Amino-2-nitropyridine |
191°C to 193°C | |
MFCD00044102 | |
124467 | |
GZBKVUGZEAJYHH-UHFFFAOYSA-N | |
2-nitropyridin-3-amine | |
83281 | |
97% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H302-H315-H319-H335
EINECSNumber : 236-260-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only